Merck
  • Electromembrane extraction combined with gas chromatography for quantification of tricyclic antidepressants in human body fluids.
Electromembrane extraction combined with gas chromatography for quantification of tricyclic antidepressants in human body fluids.
Analytica chimica acta (2012-04-17)
Saied Saeed Hosseiny Davarani, Amin Morteza Najarian, Saeed Nojavan, Mohammad-Ali Tabatabaei
ABSTRACT
Recent advances in electromembrane extraction (EME) methodology calls for effective and accessible detection methods. Using imipramine and clomipramine as model therapeutics, this proof-of-principle work combines EME with gas chromatography analysis employing a flame ionization detector (FID). The drugs were extracted from acidic aqueous sample solutions, through a supported liquid membrane (SLM) consisting of 2-nitrophenyl octyl ether (NPOE) impregnated on the walls of the hollow fiber. EME parameters, such as SLM composition, type of ion carrier, pH and the composition of donor and acceptor solutions, agitation speed, extraction voltage, and extraction time were studied in detail. Under optimized conditions, the therapeutics were effectively extracted from different matrices with recoveries ranging from 90 to 95%. The samples were preconcentrated 270-280 times prior to GC analysis. Reliable linearity was also achieved for calibration curves with a regression coefficient of at least 0.995. Detection limits and intra-day precision (n=3) were less than 0.7 ng mL(-1) and 8.5%, respectively. Finally, method was applied to determination and quantification of drugs in human plasma and urine samples and satisfactory results were achieved.
Sigma-Aldrich
Clomipramine hydrochloride, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Supelco
Clomipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.