Merck
  • High-dose, extended-interval colistin administration in critically ill patients: is this the right dosing strategy? A preliminary study.
High-dose, extended-interval colistin administration in critically ill patients: is this the right dosing strategy? A preliminary study.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America (2012-03-17)
Lidia Dalfino, Filomena Puntillo, Adriana Mosca, Rosa Monno, Maria Luigia Spada, Sara Coppolecchia, Giuseppe Miragliotta, Francesco Bruno, Nicola Brienza
ABSTRACT
Gram-negative bacteria susceptible only to colistin (COS) are emerging causes of severe nosocomial infections, reviving interest in the use of colistin. However, consensus on the most effective way to administer colistin has not yet been reached. All patients who had sepsis due to COS gram-negative bacteria or minimally susceptible gram-negative bacteria and received intravenous colistimethate sodium (CMS) were prospectively enrolled. The CMS dosing schedule was based on a loading dose of 9 MU and a 9-MU twice-daily fractioned maintenance dose, titrated on renal function. For each CMS course, clinical cure, bacteriological clearance, daily serum creatinine clearance, and estimated creatinine clearance were recorded. Twenty-eight infectious episodes due to Acinetobacter baumannii (46.4%), Klebsiella pneumoniae (46.4%), and Pseudomonas aeruginosa (7.2%) were analyzed. The main types of infection were bloodstream infection (64.3%) and ventilator-associated pneumonia (35.7%). Clinical cure was observed in 23 cases (82.1%). Acute kidney injury developed during 5 treatment courses (17.8%), did not require renal replacement therapy, and subsided within 10 days from CMS discontinuation. No correlation was found between variation in serum creatinine level (from baseline to peak) and daily and cumulative doses of CMS, and between variation in serum creatinine level (from baseline to peak) and duration of CMS treatment. Our study shows that in severe infections due to COS gram-negative bacteria, the high-dose, extended-interval CMS regimen has a high efficacy, without significant renal toxicity.
Product Description
Colistimethate sodium, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Colistin sodium methanesulfonate, ~11,500 U/mg
Sigma-Aldrich
Colistin sodium methanesulfonate, from Bacillus colistinus