Merck
  • Upregulation of heme oxygenase-1 by acteoside through ERK and PI3 K/Akt pathway confer neuroprotection against beta-amyloid-induced neurotoxicity.
Upregulation of heme oxygenase-1 by acteoside through ERK and PI3 K/Akt pathway confer neuroprotection against beta-amyloid-induced neurotoxicity.
Neurotoxicity research (2011-12-08)
Hong-Quan Wang, Yu-Xia Xu, Cui-Qing Zhu
ABSTRACT
Our previous study has shown that acteoside, an antioxidative phenylethanoid glycoside, protect against beta-amyloid (Aβ)-induced cytotoxicity in vitro. However, the precise protective mechanisms remains unclear. Heme oxygenase-1 (HO-1) is a crucial factor in the response to oxidative injury, protecting neurons against Aβ-induced injury. In the present study we examined to determine whether acteoside upregulates HO-1 expression, and thereby protects PC12 cells against Aβ-induced cell death. It was revealed that acteoside is an activator of Nrf2 and inducer of HO-1 expression. We showed that acteoside increased HO-1 expression in vitro and in vivo. Acteoside treatment resulted in nuclear translocation of the transcription factor NF-E2-related factor 2 (Nrf2). Acteoside activated both ERK and PI3 K/Akt, and treatments with the specific ERK inhibitor PD98059, the PI3 K inhibitor LY294002, and the specific Nrf2 siRNA suppressed the acteoside-induced HO-1 expression. The HO-1 inhibitor ZnPP, PD98059, and LY294002 markedly abolished the neuroprotective effect of acteoside against Aβ-induced neurotoxicity. Taken together, these results demonstrate that acteoside is an activator of Nrf2 and inducer of HO-1 expression. We also showed that acteoside increased HO-1 expression through activation of ERK and PI3 K/Akt signal pathways in vitro. Upregulation of HO-1 by acteoside may involve in the neuroprotection against Aβ-induced neurotoxicity.
Sigma-Aldrich
Verbascoside, ≥99% (HPLC)
Verbascoside, primary reference standard