Merck
  • Flow cytometry as a tool for analyzing changes in Plasmodium falciparum cell cycle following treatment with indol compounds.
Flow cytometry as a tool for analyzing changes in Plasmodium falciparum cell cycle following treatment with indol compounds.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2011-10-22)
Desirée Cigaran Schuck, Ramira Yuri Ribeiro, Arthur A Nery, Henning Ulrich, Célia R S Garcia
ABSTRACT
Melatonin and its derivatives modulate the Plasmodium falciparum and Plasmodium chabaudi cell cycle. Flow cytometry was employed together with the nucleic acid dye YOYO-1 allowing precise discrimination between mono- and multinucleated forms of P. falciparum-infected red blood cell. The use of YOYO-1 permitted excellent discrimination between uninfected and infected red blood cells as well as between early and late parasite stages. Fluorescence intensities of schizont-stage parasites were about 10-fold greater than those of ring-trophozoite form parasites. Melatonin and related indolic compounds including serotonin, N-acetyl-serotonin and tryptamine induced an increase in the percentage of multinucleated forms compared to non-treated control cultures. YOYO-1 staining of infected erythrocyte and subsequent flow cytometry analysis provides a powerful tool in malaria research for screening of bioactive compounds.
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
Tryptamine, ≥97%