- Application of a chiral metal-organic framework in enantioselective separation.
Application of a chiral metal-organic framework in enantioselective separation.
Chemical communications (Cambridge, England) (2011-10-01)
Mohan Padmanaban, Philipp Müller, Christian Lieder, Kristina Gedrich, Ronny Grünker, Volodymyr Bon, Irena Senkovska, Sandra Baumgärtner, Sabine Opelt, Silvia Paasch, Eike Brunner, Frank Glorius, Elias Klemm, Stefan Kaskel
PMID21960280
ABSTRACT
A modular approach for the synthesis of highly ordered porous and chiral auxiliary (Evans auxiliary) decorated metal-organic frameworks is developed. Our synthesis strategy, which uses known porous structures as model materials for incorporation of chirality via linker modification, can provide access to a wide range of porous materials suitable for enantioselective separation and catalysis. Chiral analogues of UMCM-1 have been synthesized and investigated for the enantioseparation of chiral compounds in the liquid phase and first promising results are reported.
MATERIALS