Merck
  • Azide-reactive liposome for chemoselective and biocompatible liposomal surface functionalization and glyco-liposomal microarray fabrication.
Azide-reactive liposome for chemoselective and biocompatible liposomal surface functionalization and glyco-liposomal microarray fabrication.
Langmuir : the ACS journal of surfaces and colloids (2011-09-21)
Yong Ma, Hailong Zhang, Valentinas Gruzdys, Xue-Long Sun
ABSTRACT
Chemically selective liposomal surface functionalization and liposomal microarray fabrication using azide-reactive liposomes are described. First, liposome carrying PEG-triphenylphosphine was prepared for Staudinger ligation with azide-containing biotin, which was conducted in PBS buffer (pH 7.4) at room temperature without a catalyst. Then, immobilization and microarray fabrication of the biotinylated liposome onto a streptavidin-modified glass slide via the specific streptavidin/biotin interaction were investigated by comparing with directly formed biotin-liposome, which was prepared by the conventional liposome formulation of lipid-biotin with all other lipid components. Next, the covalent microarray fabrication of liposome carrying triphenylphosphine onto an azide-modified glass slide and its further glyco-modification with azide-containing carbohydrate were demonstrated for glyco-liposomal microarray fabrication via Staudinger ligation. Fluorescence imaging confirmed the successful immobilization and protein binding of the intact immobilized liposomes and arrayed glyco-liposomes. The azide-reactive liposome provides a facile strategy for membrane-mimetic glyco-array fabrication, which may find important biological and biomedical applications such as studying carbohydrate-protein interactions and toxin and antibody screening.
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%