Merck
  • Stable aluminium fluoride chelates with triazacyclononane derivatives proved by X-ray crystallography and 18F-labeling study.
Stable aluminium fluoride chelates with triazacyclononane derivatives proved by X-ray crystallography and 18F-labeling study.
Chemical communications (Cambridge, England) (2011-07-22)
Dinesh Shetty, Soo Young Choi, Jae Min Jeong, Ji Youn Lee, Lathika Hoigebazar, Yun-Sang Lee, Dong Soo Lee, June-Key Chung, Myung Chul Lee, Young Keun Chung
ABSTRACT
A single crystal structure of an aluminium-fluoride complex of a model compound (NODA-benzyl) was studied to understand the co-ordination chemistry. Series of ligands with an extra carboxylic acid linker for biomolecule conjugation were studied for improved (18)F-labeling applications.
Sigma-Aldrich
Aluminum fluoride, powder, 99.8% trace metals basis