Merck
  • Application of MDPF and fluorescamine XV chiroptical properties of MDPF condensation compounds with dipeptides in situ.
Application of MDPF and fluorescamine XV chiroptical properties of MDPF condensation compounds with dipeptides in situ.
Biochemical and biophysical research communications (1983-07-29)
V Toome, B Wegrzynski
ABSTRACT
2-Methoxy-2,4-diphenyl-3(2H)-furanone (MDPF) reacts readily with the free amino group of a dipeptide to form a pyrrolinone-type chromophore with absorption maxima at 275-285 and 370-390 nm. A simple test tube procedure is described which allows in situ correlation of the absolute configuration of the NH2-terminal amino acid of a dipeptide with the chiroptical properties of its chromophoric derivative. In several cases, unexpected deviation of the chiroptical characteristics from previously established empirical rules is observed.
Sigma-Aldrich
2-Methoxy-2,4-diphenyl-3(2H)-furanone, suitable for fluorescence, ≥98.0% (HPLC)
Pricing and availability is not currently available.