Merck
  • Human plasmacytoid dendritic cells induce CD8⁺ LAG-3⁺ Foxp3⁺ CTLA-4⁺ regulatory T cells that suppress allo-reactive memory T cells.
Human plasmacytoid dendritic cells induce CD8⁺ LAG-3⁺ Foxp3⁺ CTLA-4⁺ regulatory T cells that suppress allo-reactive memory T cells.
European journal of immunology (2011-04-07)
Patrick P C Boor, Herold J Metselaar, Sarina de Jonge, Shanta Mancham, Luc J W van der Laan, Jaap Kwekkeboom
ABSTRACT
Allo-reactive memory T cells are a major barrier for induction of immunological tolerance to allografts in humans. Here, we report that stimulation of unfractionated human T cells with TLR-stimulated allogeneic plasmacytoid dendritic cells (pDCs) induces CD8(+) regulatory T cells (Tregs) that inhibit T-cell allo-responses, including those of memory T cells. CD3(+) T cells were primed for 7 days with allogeneic pDCs that had been pre-stimulated with TLR-7 or TLR-9 ligands. While the T cells proliferated and produced cytokines during the priming culture, they were profoundly hypo-responsive to re-stimulation with the same allo-antigen in a second culture. Moreover, T cells primed by pDCs exerted donor-specific suppression on allo-responses of both unfractionated and memory CD3(+) T cells. The regulatory capacity of pDC-primed T cells was confined to CD8(+) LAG-3(+) Foxp3(+) CTLA-4(+) T cells, which suppressed allogeneic T-cell responses through a CTLA-4-dependent mechanism. Induction of CD8(+) Tregs by pDCs could be partially prevented by 1-methyl tryptophan, an inhibitor of indoleamine 2,3-dioxygenase. In conclusion, stimulation of human T cells by TLR-stimulated allogeneic pDCs induces CD8(+) Tregs that inhibit allogeneic T-cell responses, including memory T cells. Donor-derived pDCs may be considered as an immunotherapeutic tool to prevent activation of the recipient allo-reactive (memory) T-cell repertoire after allogeneic transplantation.
Sigma-Aldrich
L-Tryptophan methyl ester hydrochloride, 98%