Merck
  • The structure of molten AgCl, AgI and their eutectic mixture as studied by molecular dynamics simulations of polarizable ion model potentials.
The structure of molten AgCl, AgI and their eutectic mixture as studied by molecular dynamics simulations of polarizable ion model potentials.
The Journal of chemical physics (2011-01-12)
Olga Alcaraz, Vicente Bitrián, Joaquim Trullàs
ABSTRACT
The structure of molten AgCl, AgI, and their eutectic mixture Ag(Cl(0.43)I(0.57)) is studied by means of molecular dynamics simulations of polarizable ion model potentials. The corresponding static coherent structure factors reproduce quite well the available neutron scattering data. The qualitative behavior of the simulated partial structure factors and radial distribution functions for molten AgCl and AgI is that predicted by the reverse Monte Carlo modeling of the experimental data. The AgI results are also in qualitative agreement with those calculated from ab initio molecular dynamics.
Sigma-Aldrich
Silver iodide, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Silver iodide, 99%
Pricing and availability is not currently available.