- [Secondary metabolites of mangrove endophytic fungus BL321 in the South China Sea].
[Secondary metabolites of mangrove endophytic fungus BL321 in the South China Sea].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-11-06)
Yong-Xiang Song, Huan-Ge Ma, Jun Wang, Lan Liu, Zhi-Gang She, Yong-Cheng Lin
PMID21049610
ABSTRACT
To study the secondary metabolites of mangrove endophytic fungus BL321. The compounds were isolated by chromatographic technique. The structures were identified by comprehensive physico-chemical properties and spectral methods. Five compounds were isolated and identified as 3,4a-dimethyl-2-oxo-2,4,4a,5,6,7-hexahydronaphtho[2,3-b]furan-5-carboxylic acid(1), cytochalasin C(2), cytochalasin D(3), 19,20-epoxycytochalasin C(4), ergosterol(5). Compound 1 is isolated from nature for the first time. Further more, several kinds of strong bioactive compounds were islolate from this fungus indicate that it may develop to be medical source microorganism.