Merck
  • The phenylquinazoline compound S-4893 is a non-competitive cytokinin antagonist that targets Arabidopsis cytokinin receptor CRE1 and promotes root growth in Arabidopsis and rice.
The phenylquinazoline compound S-4893 is a non-competitive cytokinin antagonist that targets Arabidopsis cytokinin receptor CRE1 and promotes root growth in Arabidopsis and rice.
Plant & cell physiology (2010-11-05)
Yuto Arata, Asako Nagasawa-Iida, Hideki Uneme, Hiroki Nakajima, Tatsuo Kakimoto, Ryo Sato
ABSTRACT
We identified two phenylquinazoline compounds in a large-scale screening for cytokinin antagonists in yeast expressing the Arabidopsis cytokinin receptor cytokinin response 1/histidine kinase 4 (CRE1). After chemical modifications, we obtained compound S-4893, which non-competitively inhibited binding of the natural ligand 2-isopentenyladenine to CRE1. S-4893 antagonized cytokinin-induced activation of the Arabidopsis response regulator 5 promoter in Arabidopsis. Importantly, S-4893 had no detectable intrinsic cytokinin agonist activity in Arabidopsis or in the transformed yeast system. Cytokinin bioassay further demonstrated that S-4893 antagonized cytokinin-induced stimulation of callus formation and inhibition of root elongation. S-4893 also promoted seminal, crown and lateral root growth in rice, suggesting that S-4893 could potentially promote root growth in a variety of agronomically important plants. We believe S-4893 will be a useful tool in functional studies of cytokinin action in a wide range of plants and a lead compound for the development of useful root growth promoters in agriculture.
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, BioReagent, suitable for plant cell culture, 1 mg/mL
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, BioReagent, suitable for plant cell culture, ≥98.5%
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥90%