Merck
  • Electrochemical biosensors based on redox carbon nanotubes prepared by noncovalent functionalization with 1,10-phenanthroline-5,6-dione.
Electrochemical biosensors based on redox carbon nanotubes prepared by noncovalent functionalization with 1,10-phenanthroline-5,6-dione.
The Analyst (2010-10-20)
Xuyan Mao, Yanhui Wu, Lanlan Xu, Xiaojuan Cao, Xiujun Cui, Liande Zhu
ABSTRACT
To improve the electrocatalytic activities of carbon nanotubes (CNT) towards the oxidation of nicotinamide adenine dinucleotide (NADH), we derive them with a redox mediator, 1,10-phenanthroline-5,6-dione (PD), by the noncovalent functionalization method. The redox carbon nanotubes (PD/CNT/GC) show excellent electrocatalytic activities towards the oxidation of NADH (catalytic reaction rate constant, k(h) = 7.26 × 10(3) M(-1) s(-1)), so the determination of NADH can be achieved with a high sensitivity of 8.77 μA mM(-1) under the potential of 0.0 V with minimal interference. We also develop an amperometric ethanol biosensor by integration of alcohol dehydrogenase (ADH) within the redox carbon nanotubes (PD/CNT/GC). The ethanol biosensor exhibits a wide linear range up to 7 mM with a lower detection limit of 0.30 mM as well as a high sensitivity of 10.85 nA mM(-1).
Sigma-Aldrich
1,10-Phenanthroline-5,6-dione, 97%
Pricing and availability is not currently available.