Merck
  • Nature of pharmacophore influences active site specificity of proteasome inhibitors.
Nature of pharmacophore influences active site specificity of proteasome inhibitors.
The Journal of biological chemistry (2010-10-13)
Michael Screen, Matthew Britton, Sondra L Downey, Martijn Verdoes, Mathias J Voges, Annet E M Blom, Paul P Geurink, Martijn D P Risseeuw, Bogdan I Florea, Wouter A van der Linden, Alexandre A Pletnev, Herman S Overkleeft, Alexei F Kisselev
ABSTRACT
Proteasomes degrade most proteins in mammalian cells and are established targets of anti-cancer drugs. The majority of proteasome inhibitors are composed of short peptides with an electrophilic functionality (pharmacophore) at the C terminus. All eukaryotic proteasomes have three types of active sites as follows: chymotrypsin-like, trypsin-like, and caspase-like. It is widely believed that active site specificity of inhibitors is determined primarily by the peptide sequence and not the pharmacophore. Here, we report that active site specificity of inhibitors can also be tuned by the chemical nature of the pharmacophore. Specifically, replacement of the epoxyketone by vinyl sulfone moieties further improves the selectivity of β5-specific inhibitors NC-005, YU-101, and PR-171 (carfilzomib). This increase in specificity is likely the basis of the decreased cytotoxicity of vinyl sulfone-based inhibitors to HeLa cells as compared with that of epoxyketone-based inhibitors.
Sigma-Aldrich
Divinyl sulfone, contains hydroquinone as inhibitor, ≥96%