Merck
  • Reactivity of alkynylzirconates towards allyl bromides: selective formation of β-allyl-zirconacyclopentadienes.
Reactivity of alkynylzirconates towards allyl bromides: selective formation of β-allyl-zirconacyclopentadienes.
Chemical communications (Cambridge, England) (2010-09-22)
Xiaoyu Yan, Yiqing Zhou, Chanjuan Xi
ABSTRACT
Reaction of alkynylzirconates with allyl bromides afforded β-allyl-zirconacyclopentadienes with high selectivity in unique reaction site.
Sigma-Aldrich
Allyl bromide, ReagentPlus®, 99%, contains ≤1000 ppm propylene oxide as stabilizer
Sigma-Aldrich
Allyl bromide, reagent grade, 97%, contains ≤1000 ppm propylene oxide as stabilizer