Merck
  • Application of modern microbiological diagnostic methods for tuberculosis in Macha, Zambia.
Application of modern microbiological diagnostic methods for tuberculosis in Macha, Zambia.
The international journal of tuberculosis and lung disease : the official journal of the International Union against Tuberculosis and Lung Disease (2010-09-08)
K E Verweij, A R Kamerik, J van Ingen, J H van Dijk, P Sikwangala, P Thuma, J L Nouwen, D van Soolingen
ABSTRACT
Macha, Zambia. To assess the benefits of auramine-O staining fluorescence microscopy and Mycobacterial Growth Indicator Tube (MGIT) liquid culture with molecular identification in tuberculosis (TB) diagnostics. One hundred patients suspected of TB were subjected to three sputum sample examinations applying Ziehl-Neelsen (ZN) and auramine-O staining and MGIT culture. Positive cultures were identified using the GenoType CM assay; cultures identified as Mycobacterium tuberculosis complex were the gold standard for a diagnosis of TB. The 100 patients produced 271 sputum samples; of these, 30 patients had positive cultures. M. tuberculosis complex bacilli were isolated in 17 (56.7%) patients, non-tuberculous mycobacteria (NTM) in 11 (36.7%) and other acid-fast bacilli in two. Forty-eight samples (17.7%) were contaminated. Auramine-O detected 16 (57.1%) patients culture-positive for mycobacteria and 12 patients with culture-proven TB, vs. respectively 8 (28.6%, P = 0.008) and 7 (41.2%, P = 0.044) for ZN. Three of eight auramine-positive/ZN-negative patients were culture-positive for NTM only. The auramine-O method significantly increases sensitivity, although the higher NTM detection rate implies that this does not in itself lead to a more accurate diagnosis of TB. MGIT culture is highly sensitive, although contamination rates were a drawback; the high frequency of NTM isolation warrants a robust identification method.
Sigma-Aldrich
Auramine O, Dye content ≥80 %, certified by the Biological Stain Commission
Sigma-Aldrich
Auramine O, Dye content 85 %, certified by the Biological Stain Commission