- Regioselective O-derivatization of quercetin via ester intermediates. An improved synthesis of rhamnetin and development of a new mitochondriotropic derivative.
Regioselective O-derivatization of quercetin via ester intermediates. An improved synthesis of rhamnetin and development of a new mitochondriotropic derivative.
The regioselective synthesis of several quercetin (3,3',4',5,7-pentahydroxy flavone) tetraesters bearing a single free OH on 5-C was achieved in good yield by proper choice of reaction conditions using common esterification procedures. Tetracetylated quercetin with the free OH on 7-C was selectively obtained instead via imidazole-promoted deacylation of the corresponding pentaester. Unambiguous structural characterization of the two isomeric tetraacetyl quercetin derivatives was obtained by combined HSQC and HMBC 2D-NMR analysis. These molecules can be used as starting materials for the regioselective synthesis of other derivatives. High yield syntheses of the natural polyphenol rhamnetin (7-O-methylquercetin) and of the new mitochondriotropic compound 7-(4-triphenylphosphoniumbutyl) quercetin iodide are reported as examples.