Merck
Orientational and translational dynamics of polyether/water solutions.
The journal of physical chemistry. B (2010-04-09)
Adam L Sturlaugson, Kendall S Fruchey, Stephen R Lynch, Sergio R Aragón, Michael D Fayer
ABSTRACT
Optical heterodyne-detected optical Kerr effect (OHD-OKE) experiments and pulsed field-gradient spin-echo NMR (PFGSE-NMR) experiments were performed to measure the rotational and translational diffusion constants of a polyether, tetraethylene glycol dimethyl ether (TEGDE), in binary mixtures with water over concentrations ranging from pure TEGDE to approaching infinite dilution. In addition, hydrodynamic calculations of the rotational and translational diffusion constants for several rigid TEGDE conformations in the neat liquid and in the infinitely dilute solution were performed to supplement the experimental data. The rotational relaxation data follow the Debye-Stokes-Einstein (DSE) equation within experimental error over the entire water concentration range. The agreement with the DSE equation indicates that there is no significant structural change of the polyether as the water content is changed. In contrast to the rotational dynamics, the translational diffusion data show a distinct deviation from Stokes-Einstein (SE) behavior. As the water content of the mixture is reduced, the translational diffusion rate decreases less rapidly than the increase in viscosity alone predicts until the water/TEGDE mole ratio of 7:1 is reached. Upon further reduction of water content, the translational diffusion tracks the viscosity. Comparison of the translational data with the rotational data and the hydrodynamic computations shows that the translational dynamics cannot be explained by a molecular shape change and that the low water fraction solutions are the ones that deviate from hydrodynamic behavior. A conjecture is presented as a possible explanation for the different behaviors of the rotational and translational dynamics.
Sigma-Aldrich
Tetraethylene glycol dimethyl ether, ≥99%