Merck
  • In vitro permeation and in vivo whitening effect of topical hesperetin microemulsion delivery system.
In vitro permeation and in vivo whitening effect of topical hesperetin microemulsion delivery system.
International journal of pharmaceutics (2010-01-12)
Yi-Hung Tsai, Ko-Feng Lee, Yaw-Bin Huang, Chi-Te Huang, Pao-Chu Wu
ABSTRACT
Hesperetin is one of the flavonoids and possess anti-inflammatory, UV-protecting and antioxidant effects. Permeation issues for topical delivery systems of such effects are occasionally problematic, and in view of the fact that microemulsions are potential carriers for transdermal delivery system, the objective of this study was to design an optimal microemulsion formulation by in vitro permeation study for hesperetin topical dosage form and determine its topical photoprotective effect and skin irritation by in vivo study. The hesperetin-loaded microemulsion showed an enhanced in vitro permeation compared to the aqueous and isopropyl myristate (IPM) suspension dosage form of hesperetin. In comparison, the effect of co-surfactant on the drug permeation capacity, propylene glycol showed highest permeation rate, followed by ethanol, glycerol and polyethylene glycol (PEG 400). Sunscreen agent padimate O, as a transdermal enhancer could increase the permeation rate of hesperetin. In case of in vivo study, the hesperetin-loaded microemulsion showed significant topical whitening effect and diminished skin irritation when compared with the non-treatment group, indicating that the hesperetin microemulsion could be used as an effective whitening agent.
Sigma-Aldrich
2-Ethylhexyl 4-(dimethylamino)benzoate, 98%