Merck
  • X-ray structural characterization of charge delocalization onto the three equivalent benzenoid rings in hexamethoxytriptycene cation radical.
X-ray structural characterization of charge delocalization onto the three equivalent benzenoid rings in hexamethoxytriptycene cation radical.
Organic letters (2009-05-29)
Vincent J Chebny, Tushar S Navale, Ruchi Shukla, Sergey V Lindeman, Rajendra Rathore
ABSTRACT
Definitive X-ray crystallographic evidence is obtained for a single hole (or a polaron) to be uniformly distributed on the three equivalent 1,2-dimethoxybenzenoid (or veratrole) rings in the hexamethoxytriptycene cation radical. This conclusion is further supported by electrochemical analysis and by the observation of an intense near-IR transition in its electronic spectrum, as well as by comparison of the spectral and electrochemical characteristics with the model compounds containing one and two dimethoxybenzene rings.
Sigma-Aldrich
1,2-Dimethoxybenzene, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dimethoxybenzene, ≥99%, FG