Merck
  • Characterization, biodistribution and small-animal SPECT of I-125-labeled c-Met binding peptide in mice bearing c-Met receptor tyrosine kinase-positive tumor xenografts.
Characterization, biodistribution and small-animal SPECT of I-125-labeled c-Met binding peptide in mice bearing c-Met receptor tyrosine kinase-positive tumor xenografts.
Nuclear medicine and biology (2009-05-09)
Eun-Mi Kim, Eun-Hye Park, Su-Jin Cheong, Chang-Moon Lee, Dong Wook Kim, Hwan-Jeong Jeong, Seok Tae Lim, Myung-Hee Sohn, Kisu Kim, Junho Chung
ABSTRACT
c-Met is a receptor tyrosine kinase involved in tumor cell growth, invasion, metastases and angiogenesis. Overexpression of c-Met is frequently observed in several tumor types. Here, we report the in vitro cell-binding properties and biodistribution and SPECT/CT imaging in glioma (U87MG) xenograft-bearing mice of (125)I-labeled c-Met-binding peptides (cMBPs) including analogs conjugated to amino acid and aliphatic carbon linkers. In vitro assays showed that the peptide without any linker and those with GGG and 8-aminooctanoic acid linkers had low cellular internalization and that IC(50) values of peptides were 1.5 microM, 65 nM and 85.3 nM, respectively. Biodistribution studies showed the GGG-containing peptide had higher tumor uptake and a higher tumor-to-blood activity concentration ratio than other receptor-binding ligands. SPECT/CT studies with a dedicated small-animal imaging system were performed in U87MG-bearing athymic mice. Although U87MG tumor xenografts could be visualized by SPECT/micro-CT using the various (125)I labeled cMBPs, image contrast and overall quality were unremarkable.
Sigma-Aldrich
8-Aminooctanoic acid, 99%