Merck
A new and facile synthesis of quinazoline-2,4(1H,3H)-diones.
Organic letters (2009-02-26)
Jiarong Li, Xian Chen, Daxin Shi, Shuling Ma, Qing Li, Qi Zhang, Jianhong Tang
ABSTRACT
A new and facile preparation of quinazoline-2,4(1H,3H)-diones was first reported which was the condensation of aromatic o-aminonitriles with DMF or N,N-diethylformamide in the presence of ZnCl(2) (0.5-10 mol %) at 190-200 degrees C in the sealed reactor.
Sigma-Aldrich
N,N-Diethylformamide, 99%