Merck
  • Discovery of highly selective and potent p38 inhibitors based on a phthalazine scaffold.
Discovery of highly selective and potent p38 inhibitors based on a phthalazine scaffold.
Journal of medicinal chemistry (2008-09-27)
Brad Herberich, Guo-Qiang Cao, Partha P Chakrabarti, James R Falsey, Liping Pettus, Robert M Rzasa, Anthony B Reed, Andreas Reichelt, Kelvin Sham, Maya Thaman, Ryan P Wurz, Shimin Xu, Dawei Zhang, Faye Hsieh, Matthew R Lee, Rashid Syed, Vivian Li, David Grosfeld, Matthew H Plant, Bradley Henkle, Lisa Sherman, Scot Middleton, Lu Min Wong, Andrew S Tasker
ABSTRACT
Investigations into the structure-activity relationships (SAR) of a series of phthalazine-based inhibitors of p38 are described. These efforts originated from quinazoline 1 and through rational design led to the development of a series of orally bioavailable, potent, and selective inhibitors. Kinase selectivity was achieved by exploiting a collection of interactions with p38alpha including close contact to Ala157, occupation of the hydrophobic gatekeeper pocket, and a residue flip with Gly110. Substitutions on the phthalazine influenced the pharmacokinetic properties, of which compound 16 displayed the most desirable profile. Oral dosing (0.03 mg/kg) of 16 in rats 1 h prior to LPS challenge gave a >50% decrease in TNFalpha production.
Sigma-Aldrich
Phthalazine, 98%