- An unusual form of canine babesiosis.
An unusual form of canine babesiosis.
The Canadian veterinary journal = La revue veterinaire canadienne (2008-04-09)
Isabel Van de Maele, Karine Savary-Bataille, Ingrid Gielen, Sylvie Daminet
PMID18390102
ABSTRACT
An Akita Inu, living in Belgium, was presented with unusual clinical manifestations of acute babesiosis that included neurological signs and pancytopenia. Diagnosis was made by identifying Babesia canis in the blood smear. Neurological signs resolved after treatment with imidocarb diproprionate. Normalization of hematological abnormalities was gradual over 5 months.