Merck
  • Allometric scaling of marbofloxacin, moxifloxacin, danofloxacin and difloxacin pharmacokinetics: a retrospective analysis.
Allometric scaling of marbofloxacin, moxifloxacin, danofloxacin and difloxacin pharmacokinetics: a retrospective analysis.
Journal of veterinary pharmacology and therapeutics (2007-09-07)
S K Cox
ABSTRACT
The purpose of this study was to examine the allometric analyses of marbofloxacin, moxifloxacin, danofloxacin and difloxacin using pharmacokinetic data from the literature. The parameters of interest (half-life, clearance and volume of distribution) were correlated across species as a function of body weight using an allometric approach (Y = aWb). Results of the allometric analysis indicated similarity between clearance and volume of distribution as they relate to body weight for all drugs. The elimination half-life was independent of body mass for all fluoroquinolones except moxifloxacin. Results of the analysis suggest that allometric scaling can be used as a tool for predicting pharmacokinetic parameters for fluoroquinolones.