Merck
  • Indole diterpene synthetic studies. Total synthesis of (+)-nodulisporic acid F and construction of the heptacyclic cores of (+)-nodulisporic acids A and B and (-)-nodulisporic acid D.
Indole diterpene synthetic studies. Total synthesis of (+)-nodulisporic acid F and construction of the heptacyclic cores of (+)-nodulisporic acids A and B and (-)-nodulisporic acid D.
The Journal of organic chemistry (2007-05-22)
Amos B Smith, Akin H Davulcu, Young Shin Cho, Kazuyuki Ohmoto, László Kürti, Haruaki Ishiyama
ABSTRACT
A first-generation strategy for construction of (+)-nodulisporic acids A (1) and B (2) is described. The strategy entails union of the eastern and western hemisphere subtargets via the indole synthesis protocol developed in our laboratory. Subsequent elaboration of rings E and F, however, revealed the considerable acid instability of the C(24) hydroxyl, thereby preventing further advancement. Nonetheless, preparation of the heptacyclic core of (+)-nodulisporic acids A and B, the total synthesis of (+)-nodulisporic acid F, the simplest member of the nodulisporic acid family, and elaboration of the heptacyclic core of (-)-nodulisporic acid D were achieved.
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%