Merck
  • Influence of polyelectrolyte on the thermosensitive property of PNIPAAm-based copolymer hydrogels.
Influence of polyelectrolyte on the thermosensitive property of PNIPAAm-based copolymer hydrogels.
Journal of materials science. Materials in medicine (2007-05-08)
Xian-Zheng Zhang, Chih-Chang Chu
ABSTRACT
A new family of poly(NIPAAm-co-2-acrylamido-2-methyl-1-propanesulfonic acid) [P(NIPAAm-co-AMPSA)] hydrogels was synthesized by incorporating negative charged AMPSA to the backbone of the PNIPAAm-based hydrogel. The effect of polyelectrolyte (i.e., PAMPSA) on the thermosensitive property of PNIPAAm hydrogels was investigated. It was found that P(NIPAAm-co-AMPSA) hydrogels exhibited unique honey-comb-like 3D porous structure having rigid cell wall as well as enhanced mechanical property. The incorporation of AMPSA into PNIPAAm backbones also led to a significant increase in swelling capability at room temperature when comparing to pure PNIPAAm hydrogels. In addition, the shrinking rate upon heating was significantly improved if the AMPSA content in P(NIPAAm-co-AMPSA) hydrogels was less than 10 wt%.
Sigma-Aldrich
Poly(2-acrylamido-2-methyl-1-propanesulfonic acid) solution, average Mw 2,000,000, 15 wt. % in H2O