Merck
  • Heteronuclear complexes of macrocyclic oxamide with co-ligands: syntheses, crystal structures, and magnetic properties.
Heteronuclear complexes of macrocyclic oxamide with co-ligands: syntheses, crystal structures, and magnetic properties.
Inorganic chemistry (2007-01-25)
Li-Na Zhu, Na Xu, Wei Zhang, Dai-Zheng Liao, Kazuyoshi Yoshimura, Ko Mibu, Zong-Hui Jiang, Shi-Ping Yan, Peng Cheng
ABSTRACT
Four heteronuclear complexes Mn(CuL)2(SCN)2 (1), {[Mn(CuL)2(mu-dca)2].2H2O}n (2), Zn(CuL)2(SCN)2 (3), and [Fe(CuL)(N3)2]2 (4) incorporating macrocyclic oxamide ligands have been synthesized and structurally characterized. L is the dianion of diethyl 5,6,7,8,15,16-hexahydro-6,7-dioxodibenzo[1,4,8,11]-tetraazacyclotetradecine-13,18-dicarboxylate, and dca is the dicyanamide. The structure of 1 or 3 consists of oxamido-bridged trinuclear [MIICuII2] molecules (for 1, M is the manganese(II) ion, and for 3, M is the zinc(II) ion). Both of them consist of 1D supramolecuar chains via pi-pi interactions. The structure of 2 also has the oxamido-bridged trinuclear [MnIICuII2] structure units and consists of 2D layers formed by the linkage of copper(II) and manganese(II) atoms via the oxamido and mu1, 5-dca bridges. Complex 4 consists of oxamido-bridged tetranuclear [FeII2CuII2] molecules and arranges in 1D chains. Different co-ligands may result in different structures in this macrocyclic oxamide system. The variable-temperature magnetic susceptibility measurements (2-300 K) of 1 and 2 both show the pronounced antiferromagnetic interactions between the copper(II) and manganese(II) ions.
Sigma-Aldrich
Oxamide, 98%