Merck
  • Tremor syndrome associated with a fungal toxin: sequelae of food contamination.
Tremor syndrome associated with a fungal toxin: sequelae of food contamination.
The Medical journal of Australia (2005-06-09)
Peter R Lewis, Michael B Donoghue, Ailsa D Hocking, Lucy Cook, Linda V Granger
ABSTRACT
We report on an elderly couple who presented with a syndrome that included severe generalised tremor and incoordination after eating soup from a damaged can. Black mould contaminating the can was subcultured and the fungus Penicillium crustosum was identified. This fungus usually produces a potent neurotoxin called penitrem A. The couple displayed symptoms consistent with penitrem A ingestion, all of which resolved fully. Penitrem A intoxication has been well documented in animals, but not in humans.
Sigma-Aldrich
Penitrem A, ≥95% (HPLC and TLC)