Merck
  • The effect of a single administration of cephapirin or cloprostenol on the reproductive performance of dairy cows with subclinical endometritis.
The effect of a single administration of cephapirin or cloprostenol on the reproductive performance of dairy cows with subclinical endometritis.
Theriogenology (2005-01-05)
R Kasimanickam, T F Duffield, R A Foster, C J Gartley, K E Leslie, J S Walton, W H Johnson
ABSTRACT
This study examined the effect of a single administration of cephapirin iu or cloprostenol im on the reproductive performance of dairy cows with subclinical endometritis. Cows (n = 228) at 20-33 days in milk (DIM) from two commercial dairy farms, determined to be normal for clinical endometritis (based on absence of abnormal uterine discharge on vaginoscopic examination) were enrolled. At enrollment, a thorough reproductive examination was performed, including rectal palpation, ultrasonography (US) and endometrial cytology (EC). The case definition for subclinical endometritis was the presence of >18% neutrophils on EC examination or fluid in uterus (FIU) on US examination. All cows were randomly assigned to receive one of three treatments: 500 mg benzathine cephapirin iu, 500 microg cloprostenol im, or control (no treatment). Reproductive performance was monitored for a minimum of 8 months after treatment. Cows with subclinical endometritis treated with cephapirin or cloprostenol had a significantly increased relative pregnancy rate compared to control [hazard ratios 1.89 (P = 0.01) and 1.70 (P = 0.05), respectively]. In conclusion, a single treatment with cephapirin or cloprostenol at 20-33 DIM significantly improved the reproductive performance of cows with subclinical endometritis.
Product Description
Supelco
Cefapirin sodium, analytical standard