Merck
  • Inhibition of nicotinoprotein (NAD+-containing) alcohol dehydrogenase by trans-4-(N,N-dimethylamino)-cinnamaldehyde binding to the active site.
Inhibition of nicotinoprotein (NAD+-containing) alcohol dehydrogenase by trans-4-(N,N-dimethylamino)-cinnamaldehyde binding to the active site.
Journal of protein chemistry (2003-12-24)
Sander R Piersma, Annika Norin, Simon de Vries, Hans Jörnvall, Johannis A Duine
ABSTRACT
Ethanol oxidation by nicotinoprotein alcohol dehydrogenase (np-ADH) from the bacterium Amycolatopsis methanolica is inhibited by trans-4-(N,N-dimethylamino)-cinnamaldehyde through direct binding to the catalytic zinc ion in a substrate-like geometry. This binding is accompanied by a characteristic red shift of the aldehyde absorbance from 398 nm to 467 nm. Np-ADH is structurally related to mammalian ADH class I, and a model of np-ADH shows how the cinnamaldehyde derivative can be accommodated in the active site of the nicotinoprotein, correlating the structural and enzymological data.