Merck
  • Anti-malondialdehyde antibodies in MRL+/+ mice treated with trichloroethene and dichloroacetyl chloride: possible role of lipid peroxidation in autoimmunity.
Anti-malondialdehyde antibodies in MRL+/+ mice treated with trichloroethene and dichloroacetyl chloride: possible role of lipid peroxidation in autoimmunity.
Toxicology and applied pharmacology (2001-02-13)
M F Khan, X Wu, G A Ansari
ABSTRACT
Trichloroethene (TCE) and one of its metabolites dichloroacetyl chloride (DCAC) are known to induce/accelerate autoimmune (AI) response in MRL+/+ mice as evident from anti-nuclear, anti-ssDNA, anti-cardiolipin, and DCAC-specific antibodies in the serum (Khan et al., Toxicol. Appl. Pharmacol. 134, 155-160, 1995). In the present study, we measured anti-malondialdehyde antibodies (AMDA) in the serum of TCE- or DCAC-treated mice in order to understand the contribution of lipid peroxidation to this AI response. Female MRL+/+ mice (5 weeks old) received ip injections of 10 mmol/kg TCE or 0.2 mmol/kg of DCAC in corn oil (100 microl) every 4(th) day for 6 weeks, while controls received an equal volume of vehicle only, and AMDA was measured in the sera of these animals by an ELISA established in our laboratory. While TCE treatment caused only marginal induction of AMDA, DCAC treatment elicited a significant AMDA response. Furthermore, a time-response study of DCAC (0.2 mmol/kg, every 4(th) day, for 2, 4, 6, or 8 weeks) showed an induction of AMDA (3/4) after 4 weeks of treatment, which was even greater at both 6 and 8 weeks of DCAC treatment (5/5). These findings were further substantiated by the presence of AMDA in systemic lupus erythematosus-prone MRL-lpr/lpr mice as early as 6 weeks of age. Presence of AMDA, as observed in this study, not only indicates increased lipid peroxidation (oxidative stress), but also suggests a putative role of oxidative stress in inflammatory autoimmune diseases.
Sigma-Aldrich
Dichloroacetyl chloride, 98%