Merck
  • A cytochrome P450 involved in the metabolism of abietane diterpenoids by Pseudomonas abietaniphila BKME-9.
A cytochrome P450 involved in the metabolism of abietane diterpenoids by Pseudomonas abietaniphila BKME-9.
Journal of bacteriology (2004-05-20)
Daryl J Smith, Vincent J J Martin, William W Mohn
ABSTRACT
Diterpenoids are naturally occurring plant compounds which have pharmaceutical properties. We have sequenced a 10.4-kbp extension of the dit cluster in Pseudomonas abietaniphila BKME-9, containing genes involved in abietane diterpenoid biodegradation. The ditQ gene was found to encode a cytochrome P450 monooxygenase, P450dit, and to be homologous to the tdtD gene of Pseudomonas diterpeniphila A19-6a. Knocking out ditQ had little effect on growth of BKME-9 on abietic acid but severely impaired growth on dehydroabietic acid (DhA) and palustric acid (PaA), increasing doubling times from 3.8 to 15 h on DhA and from 5.6 to 18.5 h on PaA. A xylE transcriptional fusion showed that transcription of ditQ was induced by a range of diterpenoids. Substrate binding assays of P450dit expressed in Escherichia coli revealed that DhA binds to the enzyme and yields a type I binding spectrum with a Kd of 0.4 microM. These results indicate that P450dit is involved in the metabolism of DhA and PaA and are consistent with its putative role in converting DhA to 7-hydroxy-DhA. Finally, an amino acid sequence identity of greater than 72% and conserved gene arrangement support the hypothesis that the dit gene cluster of P. abietaniphila BKME-9 and the tdt cluster of P. diterpeniphila A19-6a contain functional homologues.
Sigma-Aldrich
Dehydroabietic acid, ≥90% (LC/MS-ELSD)