Merck
  • Effects of a novel arginine methyltransferase inhibitor on T-helper cell cytokine production.
Effects of a novel arginine methyltransferase inhibitor on T-helper cell cytokine production.
The FEBS journal (2010-03-30)
Kevin Bonham, Saskia Hemmers, Yeon-Hee Lim, Dawn M Hill, M G Finn, Kerri A Mowen
ABSTRACT
The protein arginine methyltransferase (PRMT) family of enzymes catalyzes the transfer of methyl groups from S-adenosylmethionine to the guanidino nitrogen atom of peptidylarginine to form monomethylarginine or dimethylarginine. We created several less polar analogs of the specific PRMT inhibitor arginine methylation inhibitor-1, and one such compound was found to have improved PRMT inhibitory activity over the parent molecule. The newly identified PRMT inhibitor modulated T-helper-cell function and thus may serve as a lead for further inhibitors useful for the treatment of immune-mediated disease.
Sigma-Aldrich
Sinefungin, 95% (HPLC), powder
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Arg17) Antibody, Upstate®, from rabbit