- Elucidation of crucial structures for a catechol-based inhibitor of plasma hyaluronan-binding protein (factor VII activating protease) autoactivation.
Elucidation of crucial structures for a catechol-based inhibitor of plasma hyaluronan-binding protein (factor VII activating protease) autoactivation.
Bioscience, biotechnology, and biochemistry (2011-10-08)
Eisaku Yamamoto, Yoshikazu Kitano, Keiji Hasumi
PMID21979097
ABSTRACT
Plasma hyaluronan-binding protein (PHBP) is a serine protease the activation of which is implicated in inflammation. Previous investigations have suggested the presence of catechol-binding sites in its proenzyme form, pro-PHBP. Here we found that compounds with plural catechol groups conjugated with strong electron-withdrawing groups, such as tyrphostin AG 537 (IC(50)=18 nM), were potent inhibitors of pro-PHBP activation.