Merck
  • An endo-β-N-acetylglucosaminidase from Enterococcus faecalis V583 responsible for the hydrolysis of high-mannose and hybrid-type N-linked glycans.
An endo-β-N-acetylglucosaminidase from Enterococcus faecalis V583 responsible for the hydrolysis of high-mannose and hybrid-type N-linked glycans.
FEMS microbiology letters (2011-11-19)
Liv Anette Bøhle, Geir Mathiesen, Gustav Vaaje-Kolstad, Vincent G H Eijsink
ABSTRACT
It has been demonstrated previously that Enterococcus faecalis produces secreted endoglycosidases that enable the bacteria to remove N-linked glycans from glycoproteins. One enzyme potentially responsible for this activity is EF0114, comprising a typical GH18 endoglycosidase domain and a GH20 domain. We have analyzed the other candidate, EF2863, and show that this predicted single domain GH18 protein is an endo-β-N-acetylglucosaminidase. EF2863 hydrolyzes the glycosidic bond between two N-acetylglucosamines (GlcNAc) in N-linked glycans of the high-mannose and hybrid type, releasing the glycan and leaving one GlcNAc attached to the protein. The activity of EF2863 is similar to that of the well known deglycosylating enzyme EndoH from Streptomyces plicatus. According to the CAZy nomenclature, the enzyme is designated EfEndo18A.
Sigma-Aldrich
Endoglycosidase F1 from Elizabethkingia miricola, recombinant, expressed in E. coli, ≥16 U/mg, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase H from Streptomyces plicatus, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Endoglycosidase F3 from Elizabethkingia miricola, recombinant, expressed in E. coli, 30 U/mg