Merck
  • Determination of nine high-intensity sweeteners in various foods by high-performance liquid chromatography with mass spectrometric detection.
Determination of nine high-intensity sweeteners in various foods by high-performance liquid chromatography with mass spectrometric detection.
Analytical and bioanalytical chemistry (2011-04-06)
Agata Zygler, Andrzej Wasik, Agata Kot-Wasik, Jacek Namieśnik
ABSTRACT
An analytical procedure involving solid-phase extraction (SPE) and high-performance liquid chromatography-mass spectrometry has been developed for the determination of nine high-intensity sweeteners authorised in the EU; acesulfame-K (ACS-K), aspartame (ASP), alitame (ALI), cyclamate (CYC), dulcin (DUL), neohesperidin dihydrochalcone (NHDC), neotame (NEO), saccharin (SAC) and sucralose (SCL) in a variety of food samples (i.e. beverages, dairy and fish products). After extraction with a buffer composed of formic acid and N,N-diisopropylethylamine at pH 4.5 in ultrasonic bath, extracts were cleaned up using Strata-X 33 μm Polymeric SPE column. The analytes were separated in gradient elution mode on C(18) column and detected by mass spectrometer working with an electrospray source in negative ion mode. To confirm that analytical method is suitable for its intended use, several validation parameters, such as linearity, limits of detection and quantification, trueness and repeatibilty were evaluated. Calibration curves were linear within a studied range of concentrations (r(2) ≥ 0.999) for six investigated sweeteners (CYC, ASP, ALI, DUL, NHDC, NEO). Three compounds (ACS-K, SAC, SCL) gave non-linear response in the investigated concentration range. The method detection limits (corresponding to signal-to-noise (S/N) ratio of 3) were below 0.25 μg mL(-1) (μg g(-1)), whereas the method quantitation limits (corresponding to S/N ratio of 10) were below 2.5 μg mL(-1) (μg g(-1)). The recoveries at the tested concentrations (50%, 100% and 125% of maximum usable dose) for all sweeteners were in the range of 84.2 ÷ 106.7%, with relative standard deviations <10% regardless of the type of sample matrix (i.e. beverage, yoghurt, fish product) and the spiking level. The proposed method has been successfully applied to the determination of the nine sweeteners in drinks, yoghurts and fish products. The procedure described here is simple, accurate and precise and is suitable for routine quality control analysis of foodstuffs.
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥95% (HPLC)
Sigma-Aldrich
Neohesperidin, ≥90% (HPLC)