Merck
  • A cell therapy approach based on iPSC-derived midbrain organoids for the restoration of motor function in a Parkinson's disease mouse model.
A cell therapy approach based on iPSC-derived midbrain organoids for the restoration of motor function in a Parkinson's disease mouse model.
Heliyon (2024-01-31)
Chong-Lei Fu, Bo-Cheng Dong, Xi Jiang, Dan Li, Jun Yao
ABSTRACT
Parkinson's disease (PD) is a neurodegenerative disease characterized by the degeneration of dopaminergic (DA) neurons in the substantia nigra and loss of DA transmission in the striatum, thus making cell transplantation an effective treatment strategy. Here, we develop a cellular therapy based on induced pluripotent stem cell (iPSC)-derived midbrain organoids. By transplanting midbrain organoid cells into the striatum region of a 6-OHDA-lesioned PD mouse model, we found that the transplanted cells survived and highly efficiently differentiated into DA neurons. Further, using a dopamine sensor, we observed that the differentiated human DA neurons could efficiently release dopamine and were integrated into the neural network of the PD mice. Moreover, starting from four weeks after transplantation, the motor function of the transplanted mice could be significantly improved. Therefore, cell therapy based on iPSC-derived midbrain organoids can be a potential strategy for the clinical treatment of PD.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®