Merck
  • Rab11 activity and PtdIns(3)P turnover removes recycling cargo from endosomes.
Rab11 activity and PtdIns(3)P turnover removes recycling cargo from endosomes.
Nature chemical biology (2018-06-20)
Carlo Cosimo Campa, Jean Piero Margaria, Abhishek Derle, Marco Del Giudice, Maria Chiara De Santis, Luca Gozzelino, Francesca Copperi, Carla Bosia, Emilio Hirsch
ABSTRACT
Directional transport of recycling cargo from early endosomes (EE) to the endocytic recycling compartment (ERC) relies on phosphatidylinositol 3-phosphate (PtdIns(3)P) hydrolysis and activation of the small GTPase Rab11. However, how these events are coordinated is yet unclear. By using a novel genetically-encoded FRET biosensor for Rab11, we report that generation of endosomal PtdIns(3)P by the clathrin-binding phosphoinositide 3-kinase class 2 alpha (PI3K-C2α) controls the activation of Rab11. Active Rab11, in turn, prompts the recruitment of the phosphatidylinositol 3-phosphatase myotubularin 1 (MTM1), eventually enabling the release of recycling cargo from the EE and its delivery toward the ERC. Our findings thus define that delivery of recycling cargo toward the ERC requires spatial and sequential coupling of Rab11 activity with PtdIns(3)P turnover.
Sigma-Aldrich
Anti-MTM1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Pricing and availability is not currently available.