Merck
  • Compartmentalization of androgen receptors at endogenous genes in living cells.
Compartmentalization of androgen receptors at endogenous genes in living cells.
Nucleic acids research (2023-10-04)
Selçuk Yavuz, Hélène Kabbech, Jente van Staalduinen, Simon Linder, Wiggert A van Cappellen, Alex L Nigg, Tsion E Abraham, Johan A Slotman, Marti Quevedo, Raymond A Poot, Wilbert Zwart, Martin E van Royen, Frank G Grosveld, Ihor Smal, Adriaan B Houtsmuller
ABSTRACT
A wide range of nuclear proteins are involved in the spatio-temporal organization of the genome through diverse biological processes such as gene transcription and DNA replication. Upon stimulation by testosterone and translocation to the nucleus, multiple androgen receptors (ARs) accumulate in microscopically discernable foci which are irregularly distributed in the nucleus. Here, we investigated the formation and physical nature of these foci, by combining novel fluorescent labeling techniques to visualize a defined chromatin locus of AR-regulated genes-PTPRN2 or BANP-simultaneously with either AR foci or individual AR molecules. Quantitative colocalization analysis showed evidence of AR foci formation induced by R1881 at both PTPRN2 and BANP loci. Furthermore, single-particle tracking (SPT) revealed three distinct subdiffusive fractional Brownian motion (fBm) states: immobilized ARs were observed near the labeled genes likely as a consequence of DNA-binding, while the intermediate confined state showed a similar spatial behavior but with larger displacements, suggesting compartmentalization by liquid-liquid phase separation (LLPS), while freely mobile ARs were diffusing in the nuclear environment. All together, we show for the first time in living cells the presence of AR-regulated genes in AR foci.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF568 F(ab′)2 fragment of antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution