Merck
IL-35 and Autoimmunity: a Comprehensive Perspective.
Clinical reviews in allergy & immunology (2015-01-27)
Jinjung Choi, Patrick S C Leung, Christopher Bowlus, M Eric Gershwin
ABSTRACT
Interleukin 35 (IL-35) is the most recently identified member of the IL-12 family of cytokines and offers the potential to be a target for new therapies for autoimmune, inflammatory, and infectious diseases. Similar to other members of the IL-12 family including IL-12, IL-23, and IL-27, IL-35 is composed of a heterodimer of α and β chains, which in the case of IL-35 are the p35 and Epstein-Barr virus-induced gene 3 (EBI3) proteins. However, unlike its proinflammatory relatives, IL-35 has immunosuppressive effects that are mediated through regulatory T and B cells. Although there are limited data available regarding the role of IL-35 in human autoimmunity, several murine models of autoimmunity suggest that IL-35 may have potent effects in regulating immunoreactivity via IL-10-dependent mechanisms. We suggest that similar effects are operational in human disease and IL-35-directed therapies hold significant promise. In particular, we emphasize that IL-35 has immunosuppressive ability that are mediated via regulatory T and B cells that are IL-10 dependent. Further, although deletion of IL-35 does not result in spontaneous breach of tolerance, recombinant IL-35 can improve autoimmune responses in several experimental models.
Sigma-Aldrich
IL-35 (mouse): FC (human), recombinant, expressed in CHO cells