Merck
  • Monoclonal antibodies to desmin, the muscle-specific intermediate filament protein.
Monoclonal antibodies to desmin, the muscle-specific intermediate filament protein.
The EMBO journal (1983-01-01)
E Debus, K Weber, M Osborn
ABSTRACT
A set of monoclonal antibodies to desmin has been isolated from a fusion of mouse myeloma cells with spleen cells from mice immunized with purified porcine desmin. Eleven group I antibodies recognized desmin in the immune blot, and using defined desmin fragments the epitope has been tentatively assigned as lying between residues 325 and 372. When cell lines were tested in immunofluorescence only the human line RD and hamster BHK-21 were positive. When tissue sections were used, skeletal, cardiac, visceral and some vascular smooth muscle cells were positive. Thus, the group I antibodies appear specific for desmin and do not recognize other intermediate filament proteins. Group II monoclonals recognized not only desmin in the immune blot but also other polypeptides. The epitope of this class is located between residues 70 and 280. In immunofluorescence on cell lines and tissues, the staining patterns of group II antibodies were more complicated and demonstrate that not only other intermediate filament proteins but also additional antigenic determinants are being recognized. The group I antibodies stain, as expected from their desmin specificity, rat and human rhabdomyosarcomas and thus appear to be useful reagents in pathology.