Merck
  • Adverse outcome pathway-based analysis of liver steatosis in vitro using human liver cell lines.
Adverse outcome pathway-based analysis of liver steatosis in vitro using human liver cell lines.
STAR protocols (2023-08-24)
Mawien Karaca, Kristin Fritsche, Dajana Lichtenstein, Özlem Vural, Katrin Kreuzer, Jimmy Alarcan, Albert Braeuning, Philip Marx-Stoelting, Tewes Tralau
ABSTRACT
Here, we present an in vitro test battery to analyze chemicals for their potential to induce liver triglyceride accumulation, a hallmark of liver steatosis. We describe steps for using HepG2 and HepaRG human hepatoma cells in conjunction with a combination of several in vitro assays covering the different molecular initiating events and key events of the respective adverse outcome pathway. This protocol is suitable for assessing single substance effects as well as mixtures allowing their classification as steatotic or non-steatotic. For complete details on the use and execution of this protocol, please refer to Luckert et al. (2018),1 Lichtenstein et al. (2020),2 and Knebel et al. (2019).3.
Sigma-Aldrich
GW3965 hydrochloride, ≥98% (HPLC), powder