Merck
  • Dynamics of the Second Extracellular Loop Control Transducer Coupling of Peptide-Activated GPCRs.
Dynamics of the Second Extracellular Loop Control Transducer Coupling of Peptide-Activated GPCRs.
International journal of molecular sciences (2023-08-12)
Marcel M Wygas, Jeannette M Laugwitz, Peter Schmidt, Matthias Elgeti, Anette Kaiser
ABSTRACT
Many peptide-activated rhodopsin-like GPCRs share a β-hairpin folding motif in the extracellular loop 2 (ECL2), which interacts with the peptide ligand while at the same time being connected to transmembrane helix 3 (TM3) via a highly conserved disulfide bond. Currently, it remains unknown whether the coupling of the specifically shaped ECL2 to TM3 influences the activation of peptide-activated GPCRs. We investigated this possibility in a selection of peptide GPCRs with known structures. Most of the receptors with cysteine to alanine mutations folded like the respective wild-type and resided in the cell membrane, challenging pure folding stabilization by the disulfide bridge. G-protein signaling of the disulfide mutants was retained to a greater extent in secretin-like GPCRs than in rhodopsin-like GPCRs, while recruitment of arrestin was completely abolished in both groups, which may be linked to alterations in ligand residence time. We found a correlation between receptor activity of the neuropeptide Y2 receptor and alterations in ECL2 dynamics using engineered disulfide bridges or site-directed spin labeling and EPR spectroscopy. These data highlight the functional importance of the TM3-ECL2 link for the activation of specific signaling pathways in peptide-activated GPCRs, which might have implications for future drug discovery.
Sigma-Aldrich
Anti-OPRM1 antibody produced in rabbit, affinity isolated antibody