Merck
  • Inhibition of serum- and glucocorticoid-induced kinase 1 ameliorates hydrocephalus in preclinical models.
Inhibition of serum- and glucocorticoid-induced kinase 1 ameliorates hydrocephalus in preclinical models.
Fluids and barriers of the CNS (2023-08-19)
Alexandra Hochstetler, Hillary Smith, Makenna Reed, Louise Hulme, Paul Territo, Amanda Bedwell, Scott Persohn, Nicola Perrotti, Lucia D'Antona, Francesca Musumeci, Silvia Schenone, Bonnie L Blazer-Yost
ABSTRACT
Hydrocephalus is a pathological accumulation of cerebrospinal fluid (CSF), leading to ventriculomegaly. Hydrocephalus may be primary or secondary to traumatic brain injury, infection, or intracranial hemorrhage. Regardless of cause, current treatment involves surgery to drain the excess CSF. Importantly, there are no long-term, effective pharmaceutical treatments and this represents a clinically unmet need. Many forms of hydrocephalus involve dysregulation in water and electrolyte homeostasis, making this an attractive, druggable target. In vitro, a combination of electrophysiological and fluid flux assays was used to elucidate secretory transepithelial electrolyte and fluid flux in a human cell culture model of the choroid plexus epithelium and to determine the involvement of serum-, glucocorticoid-induced kinase 1 (SGK1). In vivo, MRI studies were performed in a genetic rat model of hydrocephalus to determine effects of inhibition of SGK1 with a novel inhibitor, SI113. In the cultured cell line, SI113 reduced secretory transepithelial electrolyte and fluid flux. In vivo, SI113 blocks the development of hydrocephalus with no effect on ventricular size of wild-type animals and no overt toxic effects. Mechanistically, the development of hydrocephalus in the rat model involves an increase in activated, phosphorylated SGK1 with no change in the total amount of SGK1. SI113 inhibits phosphorylation with no changes in total SGK1 levels in the choroid plexus epithelium. These data provide a strong preclinical basis for the use of SGK1 inhibitors in the treatment of hydrocephalus.
Sigma-Aldrich
Anti-SGK1 Antibody, Upstate®, from rabbit