Merck
  • Double DAP-seq uncovered synergistic DNA binding of interacting bZIP transcription factors.
Double DAP-seq uncovered synergistic DNA binding of interacting bZIP transcription factors.
Nature communications (2023-05-06)
Miaomiao Li, Tao Yao, Wanru Lin, Will E Hinckley, Mary Galli, Wellington Muchero, Andrea Gallavotti, Jin-Gui Chen, Shao-Shan Carol Huang
ABSTRACT
Many eukaryotic transcription factors (TF) form homodimer or heterodimer complexes to regulate gene expression. Dimerization of BASIC LEUCINE ZIPPER (bZIP) TFs are critical for their functions, but the molecular mechanism underlying the DNA binding and functional specificity of homo- versus heterodimers remains elusive. To address this gap, we present the double DNA Affinity Purification-sequencing (dDAP-seq) technique that maps heterodimer binding sites on endogenous genomic DNA. Using dDAP-seq we profile twenty pairs of C/S1 bZIP heterodimers and S1 homodimers in Arabidopsis and show that heterodimerization significantly expands the DNA binding preferences of these TFs. Analysis of dDAP-seq binding sites reveals the function of bZIP9 in abscisic acid response and the role of bZIP53 heterodimer-specific binding in seed maturation. The C/S1 heterodimers show distinct preferences for the ACGT elements recognized by plant bZIPs and motifs resembling the yeast GCN4 cis-elements. This study demonstrates the potential of dDAP-seq in deciphering the DNA binding specificities of interacting TFs that are key for combinatorial gene regulation.
Sigma-Aldrich
Anti-SBP-tag Antibody, clone 20 | MAB10764, clone 20, 1.0 mg/mL, from mouse
Pricing and availability is not currently available.