Merck
  • Succinyl-5-aminoimidazole-4-carboxamide-1-ribose 5'-Phosphate (SAICAR) Activates Pyruvate Kinase Isoform M2 (PKM2) in Its Dimeric Form.
Succinyl-5-aminoimidazole-4-carboxamide-1-ribose 5'-Phosphate (SAICAR) Activates Pyruvate Kinase Isoform M2 (PKM2) in Its Dimeric Form.
Biochemistry (2016-08-03)
Ming Yan, Srinivas Chakravarthy, Joshua M Tokuda, Lois Pollack, Gregory D Bowman, Young-Sam Lee
ABSTRACT
Human pyruvate kinase isoform M2 (PKM2) is a glycolytic enzyme isoform implicated in cancer. Malignant cancer cells have higher levels of dimeric PKM2, which is regarded as an inactive form of tetrameric pyruvate kinase. This perceived inactivity has fueled controversy about how the dimeric form of pyruvate kinase might contribute to cancer. Here we investigate enzymatic properties of PKM2(G415R), a variant derived from a cancer patient, which we show by size-exclusion chromatography and small-angle X-ray scattering to be a dimer that cannot form a tetramer in solution. Although PKM2(G415R) binds to fructose 1,6-bisphosphate (FBP), unlike the wild type this PKM2 variant shows no activation by FBP. In contrast, PKM2(G415R) is activated by succinyl-5-aminoimidazole-4-carboxamide-1-ribose 5'-phosphate (SAICAR), an endogenous metabolite that we previously showed correlates with an increased level of cell proliferation and promotes protein kinase activity of PKM2. Our results demonstrate an important and unexpected enzymatic activity of the PKM2 dimer that likely has a key role in cancer progression.
Sigma-Aldrich
SAICAR, ≥95% (HPLC)