Merck
  • Ageing in men with normal spermatogenesis alters spermatogonial dynamics and nuclear morphology in Sertoli cells.
Ageing in men with normal spermatogenesis alters spermatogonial dynamics and nuclear morphology in Sertoli cells.
Andrology (2019-06-30)
E Pohl, V Höffken, S Schlatt, S Kliesch, J Gromoll, J Wistuba
ABSTRACT
Ageing in men is believed to be associated with fertility decline and elevated risk of congenital disorders for the offspring. The previous studies also reported reduced germ and Sertoli cell numbers in older men. However, it is not clear whether ageing in men with normal spermatogenesis affects the testis and germ cell population dynamics in a way sufficient for transmitting adverse age effects to the offspring. We examined men with normal spermatogenesis at different ages concerning effects on persisting testicular cell types, that is the germ line and Sertoli cells, as these cell populations are prone to be exposed to age effects. Ageing was assessed in testicular biopsies of 32 patients assigned to three age groups: (i) 28.8 ± 2.7 years; (ii) 48.1 ± 1 years; and (iii) 70.9 ± 6.2 years, n = 8 each, with normal spermatogenesis according to the Bergmann-Kliesch score, and in a group of meiotic arrest patients (29.9 ± 3.8 years, n = 8) to decipher potential links between different germ cell types. Besides morphometry of seminiferous tubules and Sertoli cell nuclei, we investigated spermatogenic output/efficiency, and dynamics of spermatogonial populations via immunohistochemistry for MAGE A4, PCNA, CREM and quantified A-pale/A-dark spermatogonia. We found a constant spermatogenic output (CREM-positive round spermatids) in all age groups studied. In men beyond their mid-40s (group 2), we detected increased nuclear and nucleolar size in Sertoli cells, indirectly indicating an elevated protein turnover. From the 7th decade (group 3) of life onwards, testes showed increased proliferation of undifferentiated spermatogonia, decreased spermatogenic efficiency and elevated numbers of proliferating A-dark spermatogonia. Maintaining normal sperm output seems to be an intrinsic determinant of spermatogenesis. Ageing appears to affect this output and might provoke compensatory proliferation increase in A spermatogonia which, in turn, might hamper germ cell integrity.
Sigma-Aldrich
Streptavidin−Peroxidase from Streptomyces avidinii, lyophilized powder
Sigma-Aldrich
Anti-Mouse IgG (H+L)-Peroxidase antibody produced in chicken, affinity isolated antibody, lyophilized powder