Merck
  • TNFR1 Contributes to Activation-Induced Cell Death of Pathological CD4+ T Lymphocytes During Ischemic Heart Failure.
TNFR1 Contributes to Activation-Induced Cell Death of Pathological CD4+ T Lymphocytes During Ischemic Heart Failure.
JACC. Basic to translational science (2022-11-08)
Vinay Kumar, Rachel Rosenzweig, Suman Asalla, Sarita Nehra, Sumanth D Prabhu, Shyam S Bansal
ABSTRACT
CD4+ T cells turn pathological during heart failure (HF). We show that the expression of tumor necrosis factor (TNF)-α and tumor necrosis factor receptor (TNFR1) increases in HF-activated CD4+ T cells. However, the role of the TNF-α/TNFR1 axis in T-cell activation/proliferation is unknown. We show that TNFR1 neutralization during T-cell activation (ex vivo) or the loss of TNFR1 in adoptively transferred HF-activated CD4+ T cells (in vivo) augments their prosurvival and proliferative signaling. Importantly, TNFR1 neutralization does not affect CD69 expression or the pathological activity of HF-activated TNFR1-/- CD4+ T cells. These results show that during HF TNFR1 plays an important role in quelling prosurvival and proliferative signals in CD4+ T cells without altering their pathological activity.
Sigma-Aldrich
Anti-Hamster IgG (whole molecule) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution