Merck
  • Oxidative stress-induced autonomous activation of the calcium/calmodulin-dependent kinase II involves disulfide formation in the regulatory domain.
Oxidative stress-induced autonomous activation of the calcium/calmodulin-dependent kinase II involves disulfide formation in the regulatory domain.
The Journal of biological chemistry (2022-10-12)
Nathália Rocco-Machado, Lo Lai, Geumsoo Kim, Yi He, Elizabeth D Luczak, Mark E Anderson, Rodney L Levine
ABSTRACT
Calcium/calmodulin-dependent protein kinase II δ (CaMKIIδ) has a pivotal role in cardiac signaling. Constitutive and deleterious CaMKII "autonomous" activation is induced by oxidative stress, and the previously reported mechanism involves oxidation of methionine residues in the regulatory domain. Here, we demonstrate that covalent oxidation leads to a disulfide bond with Cys273 in the regulatory domain causing autonomous activity. Autonomous activation was induced by treating CaMKII with diamide or histamine chloramine, two thiol-oxidizing agents. Autonomy was reversed when the protein was incubated with DTT or thioredoxin to reduce disulfide bonds. Tryptic mapping of the activated CaMKII revealed formation of a disulfide between Cys273 and Cys290 in the regulatory domain. We determined the apparent pKa of those Cys and found that Cys273 had a low pKa while that of Cys290 was elevated. The low pKa of Cys273 facilitates oxidation of its thiol to the sulfenic acid at physiological pH. The reactive sulfenic acid then attacks the thiol of Cys290 to form the disulfide. The previously reported CaMKII mutant in which methionine residues 281 and 282 were mutated to valine (MMVV) protects mice and flies from cardiac decompensation induced by oxidative stress. Our initial hypothesis was that the MMVV mutant underwent a conformational change that prevented disulfide formation and autonomous activation. However, we found that the thiol-oxidizing agents induced autonomy in the MMVV mutant and that the mutant undergoes rapid degradation by the cell, potentially preventing accumulation of the injurious autonomous form. Together, our results highlight additional mechanistic details of CaMKII autonomous activation.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Syntide 2